Learn More
Medchemexpress LLC ART812 | 2607138-82-7 | 99.9% | C19H16ClF4N3O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13928910MG
Encompass_Preferred
ART812 is a small-molecule research inhibitor of DNA polymerase theta (Polθ) used to study microhomology-mediated end joining (MMEJ) and synthetic lethality in homologous recombination-deficient cancer models. It is orally active and characterized in biochemical and cell-based assays.
  • Potent biochemical inhibitor (IC50 7.6 nM).
  • Cell-based MMEJ inhibition (IC50 240 nM).
  • High purity solid suitable for research (99.9%).
  • Identified chemical data: CAS 2607138-82-7, formula C19H16ClF4N3O4, MW 461.79.
  • Intended for biochemical and cell-based assay applications; research use only.
Catalog No. 50-002-59324