Learn More
Cayman Chemical Artemether-d3 1mg
SDP
Supplier:  Cayman Chemical 285171 MG
Encompass_Preferred
An internal standard for the quantification of artemether by GC- or LC-MS
Catalog No. 50-294-1185