Learn More
Medchemexpress LLC Artemisic acid | 80286-58-4 | 234.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN1984100MG
Encompass_Preferred
Artemisic acid is a sesquiterpene isolated from *Artemisia annua* L. It serves as an important precursor for the synthesis of Artemisinin and exhibits various pharmacological activities. It is suitable for laboratory research in areas such as inflammation, metabolic disease, and cancer.
  • Isolated from *Artemisia annua* L.
  • Important precursor for the synthesis of Artemisinin
  • Exhibits antimalarial, antitumor, antipyretic, antibacterial, allelopathic, and anti-adipogenic effects
  • High purity of 99.74%
  • Inhibits adipogenesis in hAMSCs by reducing triglyceride accumulation and decreasing GPDH activity
  • Downregulates C/EBP δ expression
  • Mechanism involves the inhibition of JNK
  • Significantly reduces TNF-α-induced IL-6 secretion in undifferentiated hAMSCs
Catalog No. 50-004-14607