Learn More
Medchemexpress LLC Artemisinin (Standard) | 63968-64-9 | 98.6% | C₁₅H₂₂O₅ | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0094R500MG
Encompass_Preferred
Artemisinin (Standard) is the analytical standard for Artemisinin, a sesquiterpene lactone isolated from *Artemisia annua L.* plants. It functions as an anti-malarial agent and inhibits the AKT signaling pathway by decreasing pAKT. It has been shown to reduce cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis, and also possesses neuroprotective effects.
  • Analytical standard for research and analytical applications
  • Functions as an anti-malarial agent
  • Inhibits AKT signaling pathway by decreasing pAKT
  • Reduces cancer cell proliferation, migration, invasion, tumorigenesis, and metastasis
  • Possesses neuroprotective effects
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-32496