Learn More
TARGETMOL CHEMICALS INC Artesunate 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0433200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Artesunate (WR-256283) is part of the artemisinin group of drugs that treat malaria. It is a semi-synthetic derivative of artemisinin that is water-soluble and may therefore be given by injection. It is on the World Health Organization's List of Essential Medicines. Purity 99.91%
Catalog No. 50-288-0736