Learn More
Cayman Chemical Artesunate-d4 1mg
SDP
Supplier:  Cayman Chemical 287691 MG
Encompass_Preferred
An internal standard for the quantification of artesunate by GC- or LC-MS
Catalog No. 50-293-9916