Learn More
Selleck Chemical LLC Artesunate 10mg 88495-63-0 WR-256283
SDP
Supplier:  Selleck Chemical LLC S226510MG
Encompass_Preferred
Artesunate (WR-256283) is a part of the artemisinin group of agents with an IC50 of < 5 ?M for small cell lung carcinoma cell line H69. It is a potential inhibitor of STAT-3 and exhibits selective cytotoxicity of cancer cells over normal cells in vitro; A potent inhibitor of EXP1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-774-6