Learn More
Med Vet International Arthoscopic Distractor for 2.4mm (3/32") Pins, J0240XA
Supplier:  Med Vet International JOR240XA
Encompass_Preferred
External Stifle Distractor. Osteoarthritic stifles can be very tight making examination and treatment of meniscal lesions difficult. Distraction of the articular surfaces using intra-articular instruments clutters the operative field. Originally developed
Catalog No. 50-005-85401