Learn More
Chondrex Inc Arthrogen-CIA 5-Clone Cocktail Kit - 10mg
Supplier:  Chondrex Inc 53010
Encompass
Collagen antibody induced arthritis induced by Arthgogen-CIA 5 clone cocktail kit is the fastest, most consistent arthritis model to study the pathogenic roles of bacterial agents involved in RA and other autoimmune diseases. Arthritis is induced in a verity of mouse strains within 24 - 48 hours by a single injection of a sub-arthritogenic dose of the monoclonal antibody cocktail followed by (a) IP injection.
Catalog No. NC1929644