Learn More
Pickering Laboratories Inc Artificial Perspiration, GMW14334, 2-Part Alkaline, Not Stabilized, each (200 mL /bottle)
SDP
Supplier:  Pickering Laboratories Inc 17000534
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. Acid and alkaline perspiration solutions are formulated according to test procedure GMW14334 and they are designed to test chemical resistance of automotive trim materials and components.
Catalog No. 50-197-4205