Learn More
Cayman Chemical ArundIc AcId 10mg
SDP
Supplier:  Cayman Chemical 2908710 MG
Encompass_Preferred
An inhibitor of astrocyte activation; reduces increases in brain GFAP and S100β levels in a rat model of permanent focal ischemia induced by middle cerebral artery occlusion at 10 mg/kg; decreases the number of TUNEL-positive neurons, reduces infarct size, and improves motor performance in the rotarod test in the same model at 3 and 10 mg/kg; also reduces descent time and time to turn in a pole test in a mouse model of MPTP-induced Parkinson's disease at 30 mg/kg
Catalog No. 50-294-0088