Learn More
Medchemexpress LLC HY-107661 10mg Medchemexpress, Arundic Acid CAS:185517-21-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10766110MG
Encompass_Preferred
Medchemexpress, HY-107661 10mg Arundic Acid CAS:185517-21-9 Arundic acid (ONO-2506) is an astrocyte-modulating agent, which delays the expansion of cerebral infarcts by modulating the activation of astrocytes through inhibition of S-100β synthesis. Arundic acid has the potential for stroke and Alzheimer’s disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0717