Learn More
Medchemexpress LLC HY-100972 1mg Medchemexpress, ARV-771 CAS:1949837-12-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009721MG
Encompass_Preferred
Medchemexpress, HY-100972 1mg ARV-771 CAS:1949837-12-0 ARV-771 is a potent BET degrader based on PROTAC technology with Kds of 34 nM, 4.7 nM, 8.3 nM, 7.6 nM, 9.6 nM, and 7.6 nM for BRD2(1), BRD2(2), BRD3(1), BRD3(2), BRD4(1), and BRD4(2), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8643