Learn More
Cayman Chemical ArylpIperazIn 5k 50mg
SDP
Supplier:  Cayman Chemical 3813550 MG
Encompass_Preferred
A FATP1 inhibitor (IC50 = 4.6 nM for the human protein); reduces neutral and polar lipid accumulation induced by linoleic acid in MDA-MB-231 cells at 12.5 µM; decreases ATP and BODIPY-palmitate levels in CD37-/- knockout BJAB cells but not in wild-type BJAB cells at 12.5 µM; induces cell death in MDA-MB-231, MCF-7, HCC1806, and BT474 cells at 12.5 µM
Catalog No. 50-291-6571