Learn More
Apexbio Technology LLC AS 602801 848344-36-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A31895.1
Encompass_Preferred
AS 602801 (CAS 848344-36-5) is a selective small-molecule inhibitor targeting c-Jun N-terminal kinase (JNK) a kinase implicated in immune and apoptotic signaling pathways Preclinical studies indicate that AS 602801 suppresses T-cell proliferation and promotes apoptosis altering gene expression in RRMS-derived CD4 and CD8 T-cells and innate receptor expression in RRMS-associated CD11b cells Animal models show beneficial effects in autoimmune disorders and neuronal apoptosis scenarios Thus AS 602801 is investigated primarily for therapeutic potential in multiple sclerosis and fibrotic conditions
Catalog No. 50-000-63181