Learn More
Selleck Chemical LLC AS-604850 S2681-5mg
SDP
Supplier:  Selleck Chemical LLC S26815MG
Encompass_Preferred
AS-604850 is a selective ATP-competitive PI3K inhibitor with IC50 of 250 nM over 80-fold selectivity for PI3K than PI3K / and 18-fold more selective for PI3K than PI3K
Catalog No. 50-311-2532