Learn More
Medchemexpress LLC AS1842856 | 836620-48-5 | MFCD30718173 | 99.9% | 347.38 g/mol | C18H22FN3O3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY100596200MG
Encompass_Preferred
AS1842856 is a small-molecule Foxo1 inhibitor used in laboratory research to probe FOXO1-regulated pathways, including autophagy. The compound (CAS 836620-48-5) is cell-permeable, reported to have potent inhibitory activity (IC50 ≈ 30 nM), and is supplied with supporting analytical documentation for research use.
  • Potent Foxo1 inhibitor activity (IC50 ≈ 30 nM).
  • High reported purity suitable for analytical and biological experiments.
  • Available as solid and as a DMSO solution for flexible formulation.
  • Defined storage stability for powder and for solutions under controlled conditions.
  • Includes datasheet, certificate of analysis, and safety data sheet for traceability and safe handling.
  • Appropriate for in vitro studies of autophagy and FOXO1 signaling pathways.
Catalog No. 50-002-13658