Learn More
Medchemexpress LLC HY-18686 5mg Medchemexpress, AS1949490 CAS:1203680-76-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY186865MG
Encompass_Preferred
Medchemexpress, HY-18686 5mg AS1949490 CAS:1203680-76-5 AS1949490 is a potent and selective SHIP-2 (SH2 domain-containing inositol 5′ phosphatase 2) inhibitor, with an IC 50 of 620 nM. AS1949490 activated glucose metabolism via up-regulation of GLUT1 gene in L6 myotubes [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2816