Learn More
Verder Scientific Inc AS200 CONTROL 100-240V 50/60HZ
Supplier:  Verder Scientific Inc 300320001/QUOTE2600092
Encompass_Preferred
Product Features:- Up to 99 sieving programs- Low noise operation thanks to optimized control- USB connection for use of EasySieve software- Suitable for dry and wet sieving- excellent separation efficiency even with short sieving times- Efficient electromagnetic drive- 3-D throwing motion which ensures optimum use of the open sieve areaand lets the sample move equally over the whole sieving surface- Setting of sieve acceleration "g" for comparable and reproducible sievingresults worldwide- Sieve stack up to 550 mm height- Free digital adjustment of all process parameters (time, amplitude or sieveacceleration, interval)- Interval operation- Optional sieve software EasySieve for control of the shaker, easy evaluationand documentation of results- Fulfils all criteria for measuring equipment related to ISO 9001Tech Specs:- Measuring Range: 25um-25 mm- Max. batch/feed capacity:3kg- Max. number of fractions 11/23
Catalog No. NC3913676