Learn More
Hach Company AS950 Portable Compact Sampler
Supplier: Hach Company ASP.CXXXC321XX
AS950 Portable Compact Sampler-230 V-Composite 10 L PE Bottle-EU plugIncludes1 ea AS950 controller on compact portable base1 ea 230V power supply (8754500EU)1 ea Bottle kit (PC010030) includes 2 5 gallon polyethylene bottle w/cap (1918) and full bottle shut-off (8996)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)