Learn More
Hach Company AS950 Portable Compact Sampler
Supplier:  Hach Company ASP.CXXXC221XX
Encompass_Preferred
AS950 Portable Compact Sampler-115 V-Composite 10 L PE Bottle-US plugIncludes1 ea AS950 controller on compact portable base1 ea 115V power supply (8754500US)1 ea Bottle kit (PC010030) includes 2 5 gallon polyethylene bottle w/cap (1918) and full bottle shut-off (8996)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)(Battery charger 8753500US sold separately)
Catalog No. 50-002-07748
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.