Learn More
Selleck Chemical LLC Asandeutertinib E5912-100MG
SDP
Supplier:  Selleck Chemical LLC E5912100MG
Encompass_Preferred
Asandeutertinib is an orally administered ATP-competitive irreversible deuterated third-generation Epidermal Growth Factor Receptor Tyrosine Kinase Inhibitor (EGFR TKI) and a deuterium-stabilized derivative of osimertinib with the potential to treat advanced EGFR-mutated NSCLC
Catalog No. 50-000-41698