Learn More
Midwest Scientific Accuris Aspire Laboratory Aspirator, w/Pump and Bottle, 115V
SDP
Supplier:  Midwest Scientific V0020
Encompass
The Benchmark Scientific V0020 Accuris ASPIRE Laboratory Aspirator incorporates a quiet, maintenance-free pump inside a sturdy, yet compact base. The vacuum is fully adjustable, with a pressure gauge that displays the vacuum level, allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications, including removal of media from tissue culture vessels or plates, and supernatants. An ergonomic, hand held aspirating device is included, and holds standard 200 to 300ul disposable pipette tips.
Catalog No. NC1807905