Learn More
Apexbio Technology LLC ASC-J9(Synonyms: Dimethylcurcumin, GO-Y025, AR degrader enhancer ASC-J9), 10mg, CAS: 52328-98-0.
SDP
Supplier:  Apexbio Technology LLC A319010
Encompass_Preferred
ASC-J9 (CAS 52328-98-0) is a small-molecule degrader targeting androgen receptor (AR) By promoting degradation of full-length AR and splice variants such as AR3/fAR ASC-J9 inhibits androgen receptor signaling pathways involved in castration-resistant prostate cancer (CRPC) progression and metastatic behavior ASC-J9 interferes with AR-dependent transcriptional modulation and AR-independent signaling pathways notably inhibiting STAT3 phosphorylation and CCL2 chemokine secretion thus reducing macrophage infiltration and prostate cancer invasiveness Research indicates ASC-J9 may represent an alternative therapeutic strategy for CRPC treatment and metastasis prevention
Catalog No. 50-000-60859