Learn More
Medchemexpress LLC Ascaridole | 512-85-6 | 98.7% | 168.23 g/mol | C10H16O2 | 1 ML

Supplier: Medchemexpress LLC HY11849410MM 1ML
Ascaridole is a naturally occurring monoterpene peroxide used as an anthelmintic and studied for antimalarial activity. Supplied as a research-grade solution, this product is intended for laboratory use and is not for human or veterinary use.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Purity 98.65% (typical).
- Molecular weight 168.23 g/mol.
- Chemical formula C10H16O2.
- Colorless to light yellow liquid appearance.
- Intended for research applications only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.