Learn More
Aviva Systems Biology Corp ASCL2 Antibody - C-terminal region (ARP38572_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38572_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASCL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ASCL2. Alternate names: ASH2, HASH2, MASH2, bHLHa45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-284-746