Learn More
Cayman Chemical Ascorbyl TetraIsopalmItate 50g
SDP
Supplier:  Cayman Chemical 3627550 G
Encompass_Preferred
A vitamin C derivative; protects against UVB-, hydrogen peroxide-, or tert-butyl hydroperoxide-induced decreases in cell viability in HaCaT keratinocytes at 100 µM; decreases IL-1α and PGE2 production in unstimulated and UVB-stimulated HaCaT keratinocytes at 20 µM; topical application increases viable epidermis thickness, stratum corneum water content, and skin smoothness and decreases skin roughness in hairless mice
Catalog No. 50-291-1543