Learn More
Medchemexpress LLC Ascr#7 | 1139837-37-8 | 96.1% | 274.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN94415MG
Encompass_Preferred
Ascr#7 is an ascaroside, a hormone that plays a crucial role in nematode development. It is expressed during the developmental stages of nematodes and has the ability to induce the formation of long-lived, highly stress-resistant dauer larvae.
  • Functions as a nematode hormone
  • Expressed during nematode development
  • Induces formation of stress-resistant dauer larvae
Catalog No. 50-004-26436