Learn More
Medchemexpress LLC Ascr#8 | 1172120-40-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN69765MG
Encompass_Preferred
Ascr#8 is a dauer-inducing ascaroside isolated from *Caenorhabditis elegans*. It synergizes with ascr#2 and ascr#3, and strongly enhances male attraction.
  • Isolated from *Caenorhabditis elegans*
  • Synergizes with ascr#2 and ascr#3
  • Strongly enhances male attraction
  • For research use only
  • Appears as a white to off-white solid
  • Has a purity of 99.39%
Catalog No. 50-004-19051