Learn More
Medchemexpress LLC HY-N6974 5mg , Ascr#2 CAS:946524-24-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69745MG
Encompass_Preferred
HY-N6974 5mg Ascr#2 CAS: 946524-24-9 Ascr#2 is an ascaroside isolated from Caenorhabditis elegans, potently promotes dauer formation, and also acts as a potent male attractant combined with ascr#3 at low concentration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1970