Learn More
ABclonal Technology ASCT2/SLC1A5 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2315620UL
Encompass_Preferred
The SLC1A5 gene encodes a sodium-dependent neutral amino acid transporter that can act as a receptor for RD114 type D retrovirus (Larriba et al 2001 [PubMed 11781704])
Catalog No. 50-000-06156