Learn More
Echelon Biosciences Research Labs a-Secretase Substrate II , Fluorogenic - 1 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 640551MG
Encompass_Preferred
CBR 6551 / a-Secretase Substrate II , Fluorogenic - 1mg , Ac-Arg-Glu(EDANS)-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys(Dabcyl)-Arg-OH Category/Application: Alzheimer's This item is non returnable
Catalog No. 50-190-8678