Learn More
Medchemexpress LLC Asengeprast | 1001288-58-9 | MFCD28502026 | 99.3% | 351.35 | C20H17NO5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10049550MG
Encompass_Preferred
FT011 (asengeprast) is an anti-fibrotic small molecule and alkyne-containing click-chemistry reagent for research use. It reduces mRNA expression of collagen I and III, inhibits collagen synthesis, and can be conjugated via copper-catalyzed azide-alkyne cycloaddition for probe development and bioconjugation.
  • Reduces collagen I and III mRNA expression and inhibits collagen synthesis.
  • Contains an alkyne moiety enabling copper-catalyzed azide-alkyne cycloaddition for bioconjugation.
  • High purity suitable for research applications.
  • Available as a solid in multiple quantities and as a DMSO solution for convenience.
  • Intended for research use only.
Catalog No. 50-002-14729