Learn More
Santa Cruz Biotechnology ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (A-5) 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC393849
Encompass
ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (A-5) is a mouse monoclonal IgG1 κ ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 antibody, cited in 1 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 2-27 at the N-terminus of ASGPR1 of human originASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (A-5) is recommended for detection of ASGPR1 of human origin by WB, IP, IF and ELISA
Catalog No. NC3900882