Learn More
MSC Ashland Conveyor Replacement Conveyor Roller, 28 in Between Frame, 1.9 in Diameter
Supplier: MSC 66065137
This replacement conveyor roller is designed for use in conveyor systems with 28 in between frames. It features a 1.9-in diameter and a 7/16-in axle, constructed from 16 gage galvanized steel for durability.
- 1.9 in diameter
- 7/16 in axle
- Galvanized steel finish
- 16 gage construction
- For 28 in between frame systems
- SR bearing type
- Country of origin: United States