Learn More
Alkali Scientific MyFuge™ Mini Centrifuge, Green Lid, with 2 Rotors, 100-240V (US Plug)
SDP
Supplier:  Alkali Scientific C9300G
Encompass_Preferred
The MyFuge™ Mini Centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge™ is extremely easy to use. Simply close the Lid and the rotor instantly reaches 6,000 rpm. Open the Lid, and the rotor quickly decelerates to a stop for removing samples. An increased capacity, eight-position microtube rotor is included and comes installed. Also included is a second rotor for 0.2mL PCR tubes and strips which is stored (together with the rotor removal key) in a convenient comparent on the bottom of the centrifuge. When the rotor is not in use, storage in this comparent prevents loss or misplacement of the rotor.
Catalog No. 50-233-6060