Learn More
Sigma Aldrich Fine Chemicals Biosciences Asialofetuin fetal ca250MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A4781250MG
Encompass
Asialofetuin is a glycoprotein with three asparagine-linked triantennary complex carbohydrate chains and terminal N-acetylgalactosamine residues.
Catalog No. 50-290-6096