Learn More
Apexbio Technology LLC Asiatic acid 464-92-6 25mg
SDP
Supplier:  Apexbio Technology LLC N135625
Encompass_Preferred
Asiatic acid (CAS 464-92-6) is a small-molecule modulator designed to influence multiple cellular signaling pathways including NF- B MAPK and PI3K/Akt Asiatic acid exerts its biological activity primarily through inhibition of pro-inflammatory mediators and cytokine production as well as scavenging reactive oxygen species In cell-based assays asiatic acid demonstrates anticancer activity with reported IC50 values typically ranging from 10 50 M depending on the cancer cell type utilized Based on these pharmacological properties asiatic acid is investigated for research applications in wound healing inflammation neuroprotection and cancer
Catalog No. 50-000-87727