Learn More
Apexbio Technology LLC Sesamin 607-80-7 20mg
SDP
Supplier:  Apexbio Technology LLC N136220
Encompass_Preferred
Sesamin is a small-molecule inhibitor targeting delta-5 desaturase It is designed to modulate fatty acid metabolism pathways thereby regulating cellular lipid biosynthetic activity Sesamin exerts its biological activity primarily through inhibition of delta-5 desaturase In neuroscience research sesamin demonstrates neuroprotective effects by interfering with oxidative stress pathways reducing neuronal damage in various experimental models Based on these pharmacological properties sesamin holds research potential in cellular metabolic regulation neuroprotection and studies of lipid metabolism and oxidative stress mechanisms
Catalog No. 50-000-87728