Learn More
Selleck Chemical LLC Asiatic Acid 25mg 464-92-6 Dammarolic acid, Asiantic acid
SDP
Supplier:  Selleck Chemical LLC S226625MG
Encompass_Preferred
Asiatic acid (Dammarolic acid, Asiantic acid) is the aglycone of asiaticoside isolated from the plant Centella asiatica, commonly used in wound healing. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-737-4