Learn More
Cayman Chemical AsIatIcosIde 10mg
SDP
Supplier:  Cayman Chemical 1181910 MG
Encompass_Preferred
The main saponin constituent of C. asiatica that has been shown to improve wound healing including increasing collagen synthesis and elevating antioxidant levels in various normal and diabetic wound models
Catalog No. 50-295-2814