Learn More
Selleck Chemical LLC Asiaticoside S3616-5mg
SDP
Supplier:  Selleck Chemical LLC S36165MG
Encompass_Preferred
Asiaticoside (Ba 2742 BRN0078195 CCRIS8995 NSC166062 Emdecassol Madecassol) the major active principle of Centella asiatica prevents ultraviolet A-dependent photoaging by suppressing ultraviolet A-induced reactive oxygen species production It also decreases DNA binding by MITF
Catalog No. 50-310-9733