Learn More
Medchemexpress LLC Asimadoline hydrochloride | 185951-07-9 | 99.8% | 451.00 | C27H31ClN2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY107384A10MM 1ML
Encompass_Preferred
Asimadoline hydrochloride is the hydrochloride salt of asimadoline, a selective, peripherally active κ-opioid receptor agonist supplied for research use. It exhibits low blood-brain barrier permeability and has been investigated for peripheral analgesic and antinociceptive effects, including potential utility in visceral pain and irritable bowel syndrome.
  • Selective κ-opioid receptor agonist activity with low-nanomolar potency.
  • Peripherally restricted compound with low central nervous system penetration.
  • High chemical purity (99.82%).
  • Molecular weight 451.00 g/mol.
  • Available as 10 mM solution in DMSO and in solid quantities for formulation.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-41477