Learn More
Medchemexpress LLC Asimadoline hydrochloride | 185951-07-9 | MFCD28987467 | 99.8% | 451.00 | C27H31ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY107384A10MG
Encompass_Preferred
Asimadoline hydrochloride is the hydrochloride salt of asimadoline, a selective and peripherally active κ-opioid receptor agonist used as a research reagent in pharmacology and in vivo studies. It is intended for laboratory research use only and not for human or clinical use.
  • High purity suitable for analytical and in vivo research.
  • Selective κ-opioid receptor agonist activity.
  • Available in small milligram sizes for dosing flexibility.
  • Offered as solid or ready-to-use DMSO solution for convenience.
  • Characterized by CAS, molecular formula, and molecular weight data.
Catalog No. 50-002-39882