Learn More
ATCC ASK; Kidney; Atlantic Salmon (Salmo salar)
Supplier:  ATCC CRL2747
Encompass
ASK [Atlantic Salmon Kidney] is an epithelial cell line that was isolated in 1998 from the kidney of an adult, female Atlantic salmon. This cell line was deposited by J Rolland, J Winton, B Krossoey.
Catalog No. 50-238-3574