Learn More
Cayman Chemical ASK1 InhIbItr 10 25mg
SDP
Supplier:  Cayman Chemical 2851225 MG
Encompass_Preferred
An orally bioavailable inhibitor of ASK1 (IC50 = 14 nM); selective for ASK1 over ASK2 (IC50 = 510 nM) as well as MEKK1, TAK-1, IKKβ, ERK1, JNK1, p38α, GSK3β, PKCθ, and B-RAF (IC50s = >10,000 nM for all); inhibits streptozotocin-induced increases in JNK and p38 phosphorylation in INS-1 pancreatic β cells in a concentration-dependent manner
Catalog No. 50-294-0601