Learn More
Medchemexpress LLC ASN02563583 | 483283-39-2 | 99.2% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12811150MG
Encompass_Preferred
ASN02563583 is a compound that modulates the activity of the GPR17 receptor, with an IC50 value of 0.64 nM in a [35S]GTPγS binding assay. It can be used in the study of neurological diseases.
  • Modulates GPR17 receptor activity.
  • Has an IC50 value of 0.64 nM in a [35S]GTPγS binding assay.
  • Suitable for the study of neurological diseases.
  • For research use only.
Catalog No. 50-003-39221