Learn More
Medchemexpress LLC ASN03576800 | 957513-35-8 | 99.6% | 285.27 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11376110MM/1ML
Encompass_Preferred
ASN03576800 is a potent inhibitor for Ebola virus matrix protein VP40, which plays a role in viral assembly and budding processes. It functions by occupying the RNA binding region of VP40.
  • Potent inhibitor for Ebola virus matrix protein VP40.
  • Occupies the RNA binding region of VP40.
  • Intended for research use only.
Catalog No. 50-004-36528