Learn More
Medchemexpress LLC ASO 556089 sodium | 96.3% | 5414.60 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1486881MG
Encompass_Preferred
ASO 556089 sodium is a 16 nucleotide length gapmer (3-10-3) that targets the human and mouse long non-coding RNA MALAT1, with the sequence: 5'-GmCATTmCTAATAGmCAGmC-3' (mC:5-methyl cytosine, 3 bases at 3' and 5' ends:constrained ethyl; cEt). This compound is for research use only.
  • Targets PCSK9 and MALAT1 in 2D HRPTEC cultures.
  • Causes target transcriptional downregulation in HRTEC cultured on chip.
  • Inhibits MALAT1 gene expression in 2D HRPTEC culture.
  • Does not cause cytotoxicity.
Catalog No. 50-003-60155