Learn More
Medchemexpress LLC Asoxime-d4 dichlorid 500ug | 500UG
SDP
Supplier:  Medchemexpress LLC HY106901AS500UG
Encompass_Preferred
Asoxime-d4 (dichloride) is the deuterium labeled Asoxime dichloride Asoxime dichloride (HI-6) is an antagonist to acetylcholine receptors (AChRs) including the nicotinic receptor 7 nAChR Asoxime dichloride involves in modulating immunity response Asoxime dichloride (HI-6) can be used as an antigen and improves vaccination efficacy in the nervous system[1
Catalog No. 50-002-39971